Learn More
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis), Thermo Scientific Chemicals
£131.66 - £705.51
Chemical Identifiers
CAS | 108374-32-9 |
---|---|
Molecular Formula | Cl2H12N4Pt |
Molecular Weight (g/mol) | 334.11 |
MDL Number | MFCD00149947 |
InChI Key | KHCPSOMSJYAQSY-UHFFFAOYSA-L |
Synonym | platinum 4+ ion hydrate hydrochloride tetraazanide |
IUPAC Name | platinum(2+) tetraamine dichloride |
SMILES | N.N.N.N.[Cl-].[Cl-].[Pt++] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10506250
|
Thermo Scientific Acros
437005000 |
500 mg | Glass bottle |
£131.66
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10189919
|
Thermo Scientific Acros
437000050 |
5 g | Glass bottle |
£705.51
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
108374-32-9 | |
334.11 | |
KHCPSOMSJYAQSY-UHFFFAOYSA-L | |
platinum(2+) tetraamine dichloride |
Cl2H12N4Pt | |
MFCD00149947 | |
platinum 4+ ion hydrate hydrochloride tetraazanide | |
N.N.N.N.[Cl-].[Cl-].[Pt++] |
Specifications
108374-32-9 | |
99.995% | |
MFCD00149947 | |
KHCPSOMSJYAQSY-UHFFFAOYSA-L | |
platinum(2+) tetraamine dichloride | |
334.11 | |
Trace Metal Basis | |
Tetraammineplatinum(II) chloride hydrate |
White | |
Cl2H12N4Pt | |
platinum 4+ ion hydrate hydrochloride tetraazanide | |
N.N.N.N.[Cl-].[Cl-].[Pt++] | |
334.11 | |
99.995% | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
- Tetraammineplatinum(II) chloride hydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.