missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Terrific Broth (Powder/Molecular Genetics)
Enriched medium used with glycerol in cultivating recombinant strains of Escherichia coli.
Brand: Fisher BioReagents™ BP2468-2
Code : 32
Additional Details : Weight : 2.10936kg
Description
Formulation Usage: 50.8g/L
Specifications
4°C | |
Powder | |
Terrific Broth | |
Water, 94.5%; Yeast extract, 2.4%; Dipotassium phosphate, 1.3%; Trptone, 1.2%; Glycerin, 0.4%; Dihydrogen potassium phosphate, 0.2% | |
Poly Bottle |
Bacterial Culture | |
Molecular genetics | |
Microbiology Media | |
Cultivation of recombinant strains of Escherichia coli | |
2000 g |