missing translation for 'onlineSavingsMsg'
Learn More

Terephthalamidoxime, 97%, Thermo Scientific™

Product Code. 15408817
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6051-62-3
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.194
MDL Number MFCD00174328
InChI Key JUXCYRYHZGAKKG-UHFFFAOYSA-N
Synonym terephthalamidoxime, n1,n4-dihydroxyterephthalimidamide, 1,4-diamidoximobenzene, n'1,n'4-dihydroxybenzene-1,4-dicarboximidamide, terephthaldiamidoxime, 1,4-benzenediamide dioxime, n,4-benzenedicarboximidamide, benzene-1,4-dicarboxamidoxime, grwntbqzuuoolq-uhfffaoysa-n, n1,n4-dihydroxybenzene-1,4-dicarboximidamide
PubChem CID 606612
IUPAC Name N-[amino-[4-[amino(nitroso)methylidene]cyclohexa-2,5-dien-1-ylidene]methyl]hydroxylamine
SMILES C1=CC(=C(N)N=O)C=CC1=C(N)NO

Specifications

Quantity 5 g
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material Terephthalamidoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'