Learn More
Teknomatic™ Laboratory Detergent
Teknomatic is safe to use on ferrous metals, ceramics, plastics and glassware
Brand: Teknon TK300/5
Code : 04
Additional Details : CAS Number : 64-02-8 Weight : 5.85500kg
Description
Teknomatic is safe to use on ferrous metals, ceramics, plastics and glassware. The product contains a highly effective water softener to prevent the build up of scale inside the washing machine and prolongs its lifespan and efficiency. Teknomatic is biodegradable and free of enzymes, sodium hypochlorite and phosphates. This formulation is highly economical with a working strength of 0.5% and is most effective at temperatures of 55°C or above. Teknomatic is not suitable for use on non-ferrous metals or polycarbonate.- Active detergent
- Non caustic
- Non foaming
- Hard water tolerant A highly effective low foam, low residue detergent for use in automatic washing machines.
Specifications
Teknomatic Detergent | |
Use in automatic washing machines | |
5 L |
Safety and Handling
- Teknomatic
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.