missing translation for 'onlineSavingsMsg'
Learn More

TCL1B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA562747

Product Code. 15790532

  • £427.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: WQMAVHTREL LSSGQMPFSQ LPAVWQLYPG RKYRAADSSF WEIADHGQID SMEQLVLTYQ PE Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 27%.

TCL1B (T Cell Leukemia/Lymphoma 1B) is a Protein Coding gene. Diseases associated with TCL1B include T-Cell Leukemia and Lymphoma. Among its related pathways are PI3K-Akt signaling pathway. An important paralog of this gene is MTCP1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCL1B
Polyclonal
Unconjugated
TCL1B
Oncogene TCL1B; oncogene TCL-1B; SYN-1; Syncytiotrophoblast-specific protein; T-cell leukemia/lymphoma 1B; T-cell leukemia/lymphoma protein 1B; T-cell lymphoma/leukemia 1B; TCL1; TCL1/ MTCP1-like 1; TCL1/MTCP1-like protein 1; TCL1B; TML1
Rabbit
Antigen affinity chromatography
RUO
9623
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O95988
TCL1B
Recombinant protein corresponding to Human TCL1B
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only