missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14464-1-AP-20UL
This item is not returnable.
View return policy
Description
The T cell specific transcription factor TCF7 activates genes involved in immune regulation and thymocyte differentiation, and is a candidate locus for genetic susceptibility to type 1 diabetes.Specifications
TCF7 | |
Polyclonal | |
Unconjugated | |
TCF7 | |
T cell factor 1, TCF 1, TCF 7, TCF1, TCF7, Transcription factor 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21414, 6932 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P36402, Q00417 | |
TCF7 | |
TCF7 Fusion Protein Ag5792 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |