missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEAL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567536
Additional Details : Weight : 0.01000kg
Description
May be involved in transcriptional regulation. Lu X.W., Zhongguo Yi Xue Ke Xue Yuan Xue Bao 23:27-31(2001). Xu X., Submitted (MAY-2000) to the EMBL/GenBank/DDBJ databases. Ota T., Nat. Genet. 36:40-45(2004).Specifications
TCEAL4 | |
Polyclonal | |
Unconjugated | |
TCEAL4 | |
NPD017; TCEAL4; TCEA-like protein 4; transcription elongation factor A (SII)-like 4; transcription elongation factor A like 4; transcription elongation factor A protein-like 4; Transcription elongation factor S-II protein-like 4; WEX7 | |
Rabbit | |
affinity chromatography | |
RUO | |
79921 | |
-20°C | |
Liquid |
Western Blot | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96EI5 | |
TCEAL4 | |
Full length Fusion protein of human TCEAL4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |