Learn More
TBCC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557144
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MESVSCSAAA VRTGDMESQR DLSLVPERLQ RREQERQLEV ERRKQKRQNQ EVEKENSHFF VATFVRERAA VEELLERAES VERL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 69%.
Tubulin-folding protein; involved in the final step of the tubulin folding pathway.Specifications
TBCC | |
Polyclonal | |
Unconjugated | |
TBCC | |
2810055C19Rik; CFC; EGK_14912; Tbcc; tubulin folding cofactor C; tubulin-folding cofactor C; Tubulin-specific chaperone C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6903 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15814 | |
TBCC | |
Recombinant protein corresponding to Human TBCC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only