missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22124-1-AP-150UL
This item is not returnable.
View return policy
Description
The product of this gene is a microtubule-associated protein that is predomitly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.Specifications
TBC1D1 | |
Polyclonal | |
Unconjugated | |
TBC1D1 | |
KIAA1108, TBC, TBC1, TBC1 domain family member 1, TBC1D1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23216, 57915 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q60949, Q86TI0 | |
Tbc1d1 | |
TBC1D1 Fusion Protein Ag5406 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |