missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Tall Form Borosilicate Glass Beakers

Product Code. 10601672 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

These beakers offer mechanical stability, resistance to thermal shock and chemicals, and are excellent for general laboratory use.

  • With graduation and spout
  • Provided in packs of 10
  • With write-on label area
  • Acc. to DIN 12331/ISO 3819
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Diameter (Metric) Outer 95 mm
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) Outer 180 mm
Quantity 10/Pk.
Type Beaker
Graduated Yes
Scale Printed Scale
Style Tall Form
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.