Learn More
TACC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554560
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VEENLSSYSL DRRVTPASET LEDPCRTESQ HKAETPHGAE EECKAETPHG AEEECRHGGV CAPAAVATSP PGAIPKEACG GAPLQGLPGE ALGCPAGVGT PVPADGTQTL Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 27%.
TACC1 is located on 8p11 chromosomal region that is amplified in approximately 15% of all breast tumor samples. The short arm of chromosome 8 also contains FGFR1 whose expression is enhanced in most breast cancer tumors. TACC family members, TACC1, TACC2, and TACC3, map very closely to the corresponding FGFR1, FGFR2, FGFR3 genes on chromosomes 4,8, and 10. Subsequently, since they are phylogenetically related, it is proposed that TACC and FGFR have similar roles in cell growth and differentiation. Also, TACC1 contains a conserved C-terminal region as in the Drosophila homolog, D-TACC. It has been shown that D-TACC is necessary for normal spindle function, and the mammalian TACC proteins appears to interact with centrosomes and microtubules in a similar manner.Specifications
TACC3 | |
Polyclonal | |
Unconjugated | |
TACC3 | |
Aint; Arnt interacting protein; ARNT-interacting protein; C86661; ERIC1; ERIC-1; Tacc3; transforming acidic coiled coil 3; transforming acidic coiled-coil containing protein 3; transforming acidic coiled-coil-containing protein 3; transforming, acidic coiled-coil containing protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6A5 | |
TACC3 | |
Recombinant protein corresponding to Human TACC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only