Learn More
Synaptophysin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 17785-1-AP-20UL
 Ver más versiones de este producto
Código de producto. 16857764
  • £135.00 / 20µL
Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones
Descripción
Descripción
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.
Especificaciones
Especificaciones
Synaptophysin
Polyclonal
Unconjugated
SYP
SYN, synaptophysin, SYP
Rabbit
Antigen Affinity Chromatography
RUO
20977, 24804, 6855
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07825, P08247, Q62277
SYP
Synaptophysin Fusion Protein Ag11781
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Ofertas especiales
Ofertas especiales