missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptophysin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17785-1-AP-150UL
This item is not returnable.
View return policy
Description
Synaptophysin is a calcium-binding and integral membrane glycoprotein present in presynaptic vesicles in almost all neurons. Synaptophysin has four transmembrane domains and it forms a complex with dynamin at high calcium concentrations suggesting an involvement in synaptic vesicle endocytosis. It is also involved in the regulation of short-term and long-term synaptic plasticity. Synaptophysin is currently the most widely used marker for nerve terminals and for differentiating neuroendocrine tumors. Mutations in the gene can result in mental retardation, X-linked 96.Specifications
Synaptophysin | |
Polyclonal | |
Unconjugated | |
SYP | |
SYN, synaptophysin, SYP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20977, 24804, 6855 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07825, P08247, Q62277 | |
SYP | |
Synaptophysin Fusion Protein Ag11781 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |