Learn More
Synaptophysin Rabbit anti-Human, CoraLite 647, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL647-17785-100UL
Description
Synaptophysin (SYP, also known as major synaptic vesicle protein p38) is a 38-kDa integral membrane glycoprotein that regulates synaptic vesicle endocytosis. It is the most abundant synaptic vesicle protein by mass. Synaptophysin is present in neuroendocrine cells and neurons that participate in synaptic transmission. Synaptophysin is a useful marker for identification of neuroendocrine cells and neoplasms. (PMID: 3010302; 21658579)
Specifications
Synaptophysin | |
Polyclonal | |
CoraLite 647 | |
100 μL | |
6855 | |
Liquid |
Immunofluorescence | |
1000 μg/mL | |
Rabbit | |
Primary | |
Human | |
IgG |