missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Sure-Grip™ Countertop Safety Cabinet
Justrite™ Sure-Grip™ Countertop Safety Cabinet designed to hold hazardous materials like acids.
Brand: Justrite™ 86042821
Additional Details : Weight : 31.30000kg
Description
- Storage cabinet
- With one self-closing door
Specifications
432 x 559 x 432 mm | |
Blue | |
4 gal. |
349 x 470 x 349 mm | |
Adjustable | |
Self-close |