missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.5M, Chem-Lab
Ideal for titrations in laboratory applications. Sulfuric Acid, 0.5M, Chem-Lab is a dilute solution of 49.04g H2SO4 per liter of water for use in volumetric analysis.
Chemical Identifiers
CAS | 7664-93-9, 7732-18-5 |
---|---|
Molecular Formula | H2O4S |
Molecular Weight (g/mol) | 98.07 |
MDL Number | MFCD00064589 |
InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
PubChem CID | 1118 |
ChEBI | CHEBI:26836 |
IUPAC Name | sulfuric acid |
SMILES | OS(O)(=O)=O |
Description
Sulfuric acid is a highly corrosive, strong mineral acid that shows different properties depending upon its concentration. It is a pungent-ethereal, colorless to slightly yellow viscous liquid. Sulfuric acid is a diprotic acid that is hygroscopic. Concentrated sulfuric acid has strong dehydrating and oxidizing properties. Dilute sulfuric acid is a good electrolyte.Chemical Identifiers
7664-93-9, 7732-18-5 | |
98.07 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
1118 | |
sulfuric acid |
H2O4S | |
MFCD00064589 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
CHEBI:26836 | |
OS(O)(=O)=O |
Specifications
<1 | |
7664-93-9, 7732-18-5 | |
0°C | |
0.5mol/L | |
Odorless | |
H2O4S | |
Liquid | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.08g/mol |
Colorless | |
49.04g H2SO4 per 1L H2O | |
100°C | |
1.03g/mL | |
1N (±0.002, 20°C) | |
1.07 cSt | |
MFCD00064589 | |
Soluble in Water | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 |
Safety and Handling
Recommended Storage : Keep container tightly closed.
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulfuric acid 0.5 mol/l
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : H2SO4/H2O