missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sudan Black B

Histochemical stain for chromosomes

£28.14 - £106.53

Chemical Identifiers

CAS 4197-25-5
Molecular Formula C29H24N6
Molecular Weight (g/mol) 456.553
MDL Number MFCD00006919
InChI Key YCUVUDODLRLVIC-UHFFFAOYSA-N
Synonym C.I. 26150; Solvent Black 3
PubChem CID 61336
IUPAC Name (2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
SMILES CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484669
View Documents
Thermo Scientific Chemicals
J62268.09
10 g
£28.14
10g
In Stock
to see stock.
Add to basket
 
15494669
View Documents Promotion Details
Thermo Scientific Chemicals
J62268.14
25 g
£55.68
25g
In Stock
to see stock.
Add to basket
 
15404679
View Documents
Thermo Scientific Chemicals
J62268.22
100 g
£106.53
100g
In Stock
to see stock.
Add to basket
 
Description

Description

Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Histochemical stain for chromosomesSudan Black B is used as a biological stain for lipids. It is a basic black dye utilized in staining triglycerides and lipids off some lipoproteins. Further, it is used in the fingerprint enhancement as well as for detecting fats. In addition to this, it is used in the study of hematological disorders.

Solubility
Soluble in oils, fats, warm petrolatum, paraffin, phenol, ethanol, acetone, benzene, toluene and hydrocarbon. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

4197-25-5
456.553
YCUVUDODLRLVIC-UHFFFAOYSA-N
61336
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
C29H24N6
MFCD00006919
C.I. 26150; Solvent Black 3
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene

Specifications

118°C to 126°C
C29H24N6
723248
C.I. 26150; Solvent Black 3
YCUVUDODLRLVIC-UHFFFAOYSA-N
(2,2-dimethyl-1,3-dihydroperimidin-6-yl)-(4-phenyldiazenylnaphthalen-1-yl)diazene
61336
Black to Brown
Sudan Black B
4197-25-5
MFCD00006919
14,8885
Soluble in oils,fats,warm petrolatum,paraffin,phenol,ethanol,acetone,benzene,toluene and hydrocarbon. Insoluble in water.
CC1(NC2=C3C(=C(C=C2)N=NC4=CC=C(C5=CC=CC=C54)N=NC6=CC=CC=C6)C=CC=C3N1)C
456.553
456.55
Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 224-087-1

missing translation for 'rtecsNumber' : SD4431500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only