missing translation for 'onlineSavingsMsg'
Learn More

Sucrose octaacetate, 98%, Thermo Scientific Chemicals

Product Code. 10677571
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-14-7
Molecular Formula C28H38O19
Molecular Weight (g/mol) 678.59
InChI Key ZIJKGAXBCRWEOL-SAXBRCJISA-N
Synonym sucrose octaacetate, octaacetylsucrose, octa-o-acetylsucrose, d-+-sucrose octaacetate, fema no. 3038, sucrose octaacetate nf, d-+-saccharose octaacetate, 2,3,4,6,1',3',4',6'-octa-o-acetylsucrose, alpha-d-glucopyranoside, 1,3,4,6-tetra-o-acetyl-beta-d-fructofuranosyl, tetraacetate
PubChem CID 31340
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-[(2S,3S,4R,5R)-3,4-diacetyloxy-2,5-bis(acetyloxymethyl)oxolan-2-yl]oxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC2(C(C(C(O2)COC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point >78.0°C
Color Cream-White to White
Density 1.2800g/mL
Boiling Point 250.0°C (1.0 mmHg)
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Quantity 1 kg
Merck Index 15,9013
Specific Gravity 1.28
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetic acid,acetone,benzene,90g/l in ethanol
Formula Weight 678.59
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sucrose octaacetate
Show More Show Less
Product Identifier
  • Sucrose octaacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'