missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STXBP1 Mouse anti-Human, Unconjugated, Clone: 1B5B3, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | STXBP1 |
---|---|
Clone | 1B5B3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849595
|
Proteintech
67137-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16839595
|
Proteintech
67137-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The STXBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STXBP1. This antibody recognizes human, mouse, rat, pig antigen. The STXBP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
STXBP1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
6812 | |
Primary |
1B5B3 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse, Rat, Pig | |
IgG2a |