Learn More
Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™
£15.87 - £467.33
Chemical Identifiers
CAS | 7783-48-4 |
---|---|
Molecular Formula | F2Sr |
Molecular Weight (g/mol) | 125.62 |
MDL Number | MFCD00011251 |
InChI Key | FVRNDBHWWSPNOM-UHFFFAOYSA-L |
Synonym | strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g |
PubChem CID | 82210 |
SMILES | [F-].[F-].[Sr++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337047
|
Thermo Scientific Alfa Aesar
010878.02 |
0.5 g | N/A | ||||||
11327047
|
Thermo Scientific Alfa Aesar
010878.06 |
5 g |
£48.64
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317047
|
Thermo Scientific Alfa Aesar
010878.14 |
25 g |
£165.83
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347047
|
Thermo Scientific Alfa Aesar
010878.22 |
100 g |
£467.33
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Strontium fluoride is used as an optical coating on lenses, and as a thermoluminescent dosimeter crystal. It also finds use as a water insoluble strontium source for use in oxygen-sensitive applications such as metal production. It serves both as a source of strontium and fluoride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-48-4 | |
125.62 | |
FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
82210 |
F2Sr | |
MFCD00011251 | |
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
[F-].[F-].[Sr++] |
Specifications
7783-48-4 | |
99.99%, metal basis | |
MFCD00011251 | |
FVRNDBHWWSPNOM-UHFFFAOYSA-L | |
strontium;difluoride | |
82210 | |
Strontium Fluoride |
Powder | |
F2Sr | |
strontium difluoride, unii-efy8gjs81z, strontium fluoride srf2, efy8gjs81z, strontium 2+ ion difluoride, fluoride, sr, fluoride, strontium 2+ difluoride, beta-bromo isovaleric acid, ksc379a4r, strontium fluoride 250g | |
[F-].[F-].[Sr++] | |
125.62 | |
125.62 |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
Irritating to eyes, respiratory system and skin.
missing translation for 'einecsNumber' : 232-000-3
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.