missing translation for 'onlineSavingsMsg'
Learn More

Stanhope Seta Strip for copper corrosion tests 76mm x 12.5 x 2.4mm

Brand:  Stanhope Seta 11550-0

Strip for copper corrosion tests 76mm x 12.5 x 2.4mm

Code : 50

Additional Details : Weight : 5.00000kg

Product Code. 12961448

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers