Learn More
IBA Lifesciences Streptavidin
GREENER_CHOICE
Streptavidin is a widely used tool for immobilization and assays of biotinylated proteins or other biotin conjugated molecules.
Brand:  IBA Lifesciences 2-0203-010
 View more versions of this product
Product Code. 12863186
  • £292.00 / 10mg
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Streptavidin is a tetrameric protein composed of identic subunits. Each subunit specifically binds one biotin molecule with fM affinity, which makes it one of the strongest non-covalent interactions known. The preparation contains an N- and C-terminal shortened variant (core streptavidin) with improved properties regarding homogeneity, solubility, resistance towards proteolytic degradation, and accessibility of the biotin binding pocket as compared to native streptavidin.
The streptavidin:biotin system is widely used for immobilization and detection of biotinylated molecules, like proteins and nucleic acids. IBA Lifesciences provides bulk amounts of lyophilized streptavidin in reliable and high quality at competitive prices to generate streptavidin-coated surfaces (microplates, SPR chips, beads) or detection reagents conjugated with, e.g., fluorescent dyes.
Please note that streptavidin is not applicable for detection, immobilization, and purification of Strep-tag™II and Twin-Strep-tag™ fusion proteins, since the binding affinity for both tags is too low.
Specifications
Specifications
13.3 kDa/subunit
10 mg
Recombinant, expression in E. coli
>95 %