missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences Strep-Tactin™ 4Flow™ high capacity FPLC column

Strep-Tactin™ 4Flow™ high capacity FPLC column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins

£124.00 - £359.00

Specifications

Binding Capacity 20 mg/mL resin
Description 4% agarose, crosslinked
Particle Size 50 to 150 μm
Form Resin Slurry
Quantity 5 mL
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
30117607
View Documents
IBA Lifesciences
2-1258-001
5 mL
£359.00
5mL
Please to purchase this item. Need a web account? Register with us today!
30117608
View Documents
IBA Lifesciences
2-1257-001
1 mL
£124.00
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Strep-Tactin™ 4Flow™ high capacity FPLC columns are designed for purifying Strep-tag™II and Twin-Strep-tag™ proteins via affinity chromatography in automated protein purification with HPLC/FPLC devices such as Äkta™ systems (Cytiva).
  • Recommended for all protein classes due to the broad pH compatibility, excellent pressure stability, outstanding target purity (>95%), and high binding affinity
  • 4% crosslinked agarose beads coupled to Strep-Tactin™ for straightforward purification of proteins with Strep-tag™II or a Twin-Strep-tag™
  • Multiple regeneration cycles possible without loss of binding capacity
  • Ideal alternative for proteins that are difficult to elute from Strep-Tactin™XT 4Flow™ resins
  • Biotinylated proteins also bind to Strep-Tactin™ 4Flow™ high capacity and need to be blocked using BioLock
Specifications

Specifications

20 mg/mL resin
50 to 150 μm
5 mL
Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Affinity chromatography equipment
Agarose
4% agarose, crosslinked
Resin Slurry
Tetrameric protein
Column
Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.