missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler
These disposable tubes can be immersed absolutely vertically into the container.
Brand: Buerkle™ 5305-1024
This item is not returnable.
View return policy
Description
- Manufactured from either polyethylene or polyamide plastic
- Polyethylene tubes used for general sampling, polyamide tubes used for flammable liquids in VbF Hazard Category AI (electrically conducting)
- Diameter of 6 x 8mm
- Not autoclavable
Specifications
Suction Tube | |
Polyamide | |
Black | |
0.8 kg | |
220 cm |
Polyamide | |
Straight | |
For Flammable Liquids in VbF Hazard Category AI (Electrically Conducting) | |
6 x 8 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ Straight Suction Tube for Buerkle™ ProfiSampler > Black; Length: 220cm; Quantity: 10
Spot an opportunity for improvement?Share a Content Correction