missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ SteriPlast™ Blue Detectable Scoops
For sampling food, animal feed and pharmaceutical industries. Buerkle™ SteriPlast™ Blue Detectable Scoops are ideal for powders, granulates and pastes and can be detected by color scanners, metal detectors and X rays.
Brand: Buerkle™ 5378-2020
145.63 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Use of blue tools helps to meet requirements of HACCP hazard analysis and assessment in reference to foreign objects
- This is important criterion in food processing and production, and in animal feed and pharmaceutical industry
- Can be used as part of HACCP/IFS foreign object management
- Individually packaged for disposable use and sterilised by gamma rays
- Production, assembly and packaging according to clean room class 7
- Can be detected by colour scanners, metal detectors or X-rays
Compliance
EU food requirements
Specifications
Sampler | |
Not Autoclavable | |
2000 mL | |
Sterile | |
Complies with EU food rrequirements and FDA regulations |
Polystyrene | |
67.63 oz. | |
Single-use | |
Blue |