missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Step stool, foldable
Brand: Manutan A353027
Code : Z3
Additional Details : Weight : 0.01000kg
Description
Foldable step stool ideal for maintenance activities. Slip resistant surfaces and foot pads. Stands upright when folded. Easy to clean, carry and store.- Load capacity : 150kg
- Opened H x W x D : 630mm x 486mm x 530mm
- Folded H x W x D : 635mm x 486mm x 140mm