missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Stationery cupboard with 3 shelves, light grey, 1800mm x 900mm x 420mm
Brand: Manutan A334385
Code : Z3
Additional Details : Weight : 1.00000kg
Description
A strong and rigid construction around a welded frame. Supplied complete with adjustable black lateral filing shelves. Underside of shelf rails set at 330mm centres to accept most popular brands of lateral pockets. Finished in textured powder paint which is tough, durable, scratch-resistant and environmentally friendly.- Light grey
- Dimensions H x W x D : 1800mm x 900mm x 420mm
- 3 shelves