missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Stainless Steel Screen

The Stainless Steel Screen is a replacement component part, a stainless steel support, for the Wheaton™ 90 mm Filtration Assembly.

Brand:  Wheaton™ 419471

Product Code. 12411591

This item is not returnable. View return policy

Description

Description

  • Replacement part for 90 mm filtration assembly
  • Stainless steel
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wheaton™ 90 mm Filtration Assembly
Stainless Steel
1 L Funnel with Screw Collar, Glass Support Base, Filter Support, No. 8 Silicone Stopper
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.