Learn More
Sampling Systems™ Stainless Steel PharmaBottles™
Sampling Systems™ Stainless Steel PharmaBottles™ are ideal for use with powders and liquids.
£274.00 - £280.00
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
15839721
|
Sampling Systems™
A846-1000 |
1000 mL | 102 mm | 178 mm |
£280.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15849721
|
Sampling Systems™
A846-500 |
500 mL | 83 mm | 154 mm |
£274.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Multi-purpose containers
- 316L Stainless Steel body and lid
- Robust construction
- Crevice free interior
- All welds ground and polished
- Complete with screw on lid
- Silicone (FDA acceptable) gasket in lid
Specifications
Stainless Steel (316L) | |
Threaded Cap |