Learn More
Sampling Systems™ Stainless Steel Ladles
These ladles are made from hygienic stainless steel and are ideal for sampling liquid products.
Brand:  Sampling Systems™ A324-120
 View more versions of this product
Product Code. 12967955
  • £18.55 / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Satin finish
  • Crevice-free construction so they can be easily cleaned between batches
  • Range of sizes available
Specifications
Specifications
Ladle
120 mL
345 mm
Stainless Steel (AISI 304)
Sampling liquid products