missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Stainless steel keylinks, 800kg break load, standard aperture
Brand: Manutan A339866
This item is not returnable.
View return policy
Description
Stainless steel, supplied in packs of two. Conform to AISI 316 standards.- Maximum capacity: 12500kg
- Aperture: 4.0mm
- Maximum load : 800kg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction