Learn More
Thermo Scientific™ Stainless-Steel Disposable Resin
Refill stainless-steel ion exchanges with this disposable, mixed-bed resin featuring advanced, regeneration service.
Brand: Thermo Scientific™ 50155538
Code : A0
Additional Details : Weight : 0.01000kg
Description
Use Thermo Scientific™ Disposable Resin to refill stainless steel ion exchangers. The mixed-bed resins have documented quality monitoring and reliably function due to an advanced regeneration service. Each regenerated batch is monitored for both consistent capacity and quality.
This service is only available in Germany.
Specifications
Resin 2 x 12 liters disposable | |
Ion Exchange | |
Mixed bed deionizer DI 2800 (part number 02.1500) |
Stainless Steel | |
Mixed Bed | |
Disposable Resin |
This service is only available in Germany.