Learn More
Gilson™ Stainless Steel Connectors
Metal sleeve connectors for small bore tubing
Brand: Gilson™ F117985
This item is not returnable.
View return policy
Description
- Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
- PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications
Connector | |
Stainless Steel | |
Metal sleeve |
0.6 mm | |
5 Set |