missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Stack and nest storage bin, blue, NXT6, 21.0 litre
Brand: Manutan A344413
Code : Z3
Additional Details : Weight : 1.00000kg
Description
The ultimate space-saving storage containers that stack and nest. Resistant to most chemicals, including oil and acids. Temperature range from -15°C to +50°C (dependant on use). Manufactured from polypropylene. Lifting handle on larger bins. Eco-white - non-pigmented colour, offering the same features and quality as red and blue for less money. Supplied with labels. Optional dividers run back to front, or left to right dependent on bin size.- Type: A6
- Blue
- Dimensions (H x W x D): 180mm x 470mm x 385mm