missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567915
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
The polyamines putrescine, spermine, and spermidine are ubiquitous polycationic mediators of cell growth and differentiation. SRM synthase is one of four enzymes in the polyamine-biosynthetic pathway and carries out the final step of spermidine biosynthesis. This enzyme catalyzes the conversion of putrescine to spermidine using decarboxylated S-adenosylmethionine as the cofactor.Specifications
SRM | |
Polyclonal | |
Unconjugated | |
SRM | |
AA407669; PAPT; Putrescine aminopropyltransferase; SpdST; SPDSY; Spermidine synthase; spermidine synthase-1; SPS1; Srm; SRML1 | |
Rabbit | |
affinity chromatography | |
RUO | |
20810, 6723, 84596 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.02% sodium azide; pH 7.3 | |
P19623, Q64674 | |
SRM | |
Synthesized peptide derived from human SRM. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |