missing translation for 'onlineSavingsMsg'
Learn More

Salter Balance 5,000g spring tubular case x 100g

Brand:  Salter SABN04100010014

Balance 5,000g spring tubular case x 100g

Code : Z1

Additional Details : Weight : 0.09776kg

Product Code. 12767056

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.