Learn More
Splunc2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560583
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VDLGVLQKSS AWQLAKQKAQ EAEKLLNNVI SKLLPTNTDI FGLKISNSLI LDVKAEPIDD GKGLNLSFPV TANVTVAGPI IGQII Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 28%.
SPLUNC2 is one member of PLUNC family, the gene undergoes alternative splicing using two 5' non-coding exons, suggesting that the gene is regulated by alternative promoters.multiple SPLUNC2 isoforms are found in the oral cavity and suggest that these proteins may be differentially regulated in distinct tissues where they may function in the innate immune response. Mucin plugs, mucous and intermediate cells of mucoepidermoid carcinomas were positive for LPLUNC1 and SPLUNC2, but areas composed of epidermoid and clear cells were negative for all PLUNCs. Papillary cystadenocarcinoma was positive for all PLUNCs.Specifications
Splunc2 | |
Polyclonal | |
Unconjugated | |
BPIFA2 | |
bA49G10.1; BPI fold containing family A member 2; BPI fold containing family A, member 2; BPI fold containing family A, member 2E; BPI fold-containing family A member 2; BPIFA2; Bpifa2e; C20orf70; mSplunc2; parotid secretory protein; PSP; short palate; short palate, lung and nasal epithelium carcinoma-associated protein 2; SPLUNC2; UNQ510/PRO1025 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96DR5 | |
BPIFA2 | |
Recombinant protein corresponding to Human Splunc2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only