Learn More
SPIDR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565304
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PALCYILTAHP NLGQIDIIDE DPIYKLYQPP VTRCLRDILQ MNDLGTRCSF YATVIYQKPQ LKSLLLLEQR EIWLLVTDVT LQTK Highest antigen sequence identity to the following orthologs - mouse 72%, rat 67%.
Plays a role in DNA double-strand break (DBS) repair via homologous recombination (HR). Serves as a scaffolding protein that helps to promote the recruitment of DNA-processing enzymes like the helicase BLM and recombinase RAD51 to site of DNA damage, and hence contributes to maintain genomic integrity.Specifications
SPIDR | |
Polyclonal | |
Unconjugated | |
SPIDR | |
DNA repair-scaffolding protein; k0146; KIAA0146; Scaffolding protein involved in DNA repair; SPIDR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23514 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14159 | |
SPIDR | |
Recombinant Human SPIDR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only