Learn More
SPDYC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558963
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPLVTTQVEL GGCSRQGGGN GFLRFRQHQE VQAFLSLLED SFVQEFLSKD PCFQISDKY Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 28%.
Promotes progression through the cell cycle via binding and activation of CDK1 and CDK2. Involved in the spindle-assembly checkpoint. Required for recruitment of MAD2L1, BUBR1 and BUB1 to kinetochores. Required for the correct localization of the active form of Aurora B in prometaphase.Specifications
SPDYC | |
Polyclonal | |
Unconjugated | |
SPDYC | |
hSpy/Ringo C; rapid inducer of G2/M progression in oocytes C; RINGO C; Ringo2; RINGOC; SPDYC; speedy; speedy C; speedy homolog C; speedy protein C; speedy/RINGO cell cycle regulator family member C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
387778 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5MJ68 | |
SPDYC | |
Recombinant protein corresponding to Human SPDYC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only