Learn More
SPATS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565083
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: SPSMLTGNSPR GCRLPSISST TCGRQLEKVP EKRDSGMTEV ERTYSANCSD FLESKGCFAN TTPSGKSVSS SSS Highest antigen sequence identity to the following orthologs - mouse 28%, rat 28%.
mRNA is expressed during spermatogenesis with highest expression in first meiotic prophase; contains a serine stretch and may be phosphorylated.Specifications
SPATS1 | |
Polyclonal | |
Unconjugated | |
Spats1 | |
1700011H05Rik; 4933400B06Rik; Daip2; DDIP; Dvl2-DEP domain interacting protein; dvl2-DEP domain-interacting protein; SPATA8; SPATS1; spermatogenesis associated serine rich 1; spermatogenesis associated, serine-rich 1; spermatogenesis-associated serine-rich protein 1; SRSP1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q496A3 | |
Spats1 | |
Recombinant Human SPATS1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only