Learn More
SPAG8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564354
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PGSGPGHGSG SHPGPASGPG PDTGPDSELS PCIPPGFRNL VADRVPNYTS WSQHCPWEPQ KQPPWEFLQV LEPGARGLWK P Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 34%.
The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein encoded by this gene is recognized by sperm agglutinating antibodies from an infertile woman. This protein is localized in germ cells of the testis at all stages of spermatogenesis and is localized to the acrosomal region of mature spermatozoa. Alternatively spliced variants that encode different protein isoforms have been described but the full-length sequences of only two have been determined.Specifications
SPAG8 | |
Polyclonal | |
Unconjugated | |
SPAG8 | |
BS-84; CILD28; CT142; HSD-1; hSMP-1; MH-SPAG8; SMP1; SMP-1; SPAG3; Spag8; sperm associated antigen 8; sperm associated antigen 8-like protein; sperm membrane protein 1; Sperm membrane protein BS-84; sperm-associated antigen 8; testicular tissue protein Li 177 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26206 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99932 | |
SPAG8 | |
Recombinant protein corresponding to Human SPAG8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only