Learn More
SOWAHB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557829
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASRVNVRDSS GKKPWQYLTS NTSGEIWQLL GAPRGKPIFP VYPLVGSSSP TRKAKSKEIS RSVTRKTSFA ALLKSQHNKW KLANQYEKFH SPR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 82%.
SOWAHB Polyclonal antibody specifically detects SOWAHB in Human samples. It is validated for Immunohistochemistry.Specifications
SOWAHB | |
Polyclonal | |
Unconjugated | |
SOWAHB | |
ANKRD56; ankyrin repeat domain 56; ankyrin repeat domain-containing protein 56; Ankyrin repeat domain-containing protein SOWAHB; protein sosondowah homolog B; sosondowah ankyrin repeat domain family member B; SOWAHB | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
345079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NEL2 | |
SOWAHB | |
Recombinant protein corresponding to Human SOWAHB | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only