missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Sorvall™ Microtube Adapters
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ adapters.
Brand: Thermo Scientific™ 76003750
193.80 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.
Ordering Information:
Sold in packs of 24
Specifications
0.2mL microtubes | |
Gray | |
24 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction