missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan sesquioleate, Thermo Scientific Chemicals

An emulsifier

£35.48 - £63.30

Chemical Identifiers

CAS 8007-43-0
Molecular Formula C42H76O7
MDL Number MFCD00151568
Synonym Arlacel™ 83; Span™ 83
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444979
View Documents
Thermo Scientific Alfa Aesar
J62648.AP
500 mL
£35.48
500mL
Please to purchase this item. Need a web account? Register with us today!
15454979
View Documents
Thermo Scientific Alfa Aesar
J62648.K2
1 L
£63.30
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel 'coated' after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan sesquioleate is an additive found in food, veterinary products, and household items. Sorbitan esters being nonionic surface agents are used as excipients in veterinary medicinal products. Emollient ester of this compound offers a smooth feel ‘coated‘ after-feel to the skin and is used in cosmetics. It acts as a emulsification and lubricating agent.

Solubility
Insoluble in cold water, hot water. soluble in mineral oil and ethyl acetate.

Notes
Keep container tightly closed. Keep container in a cool, well-ventilated area and protect from moisture. 
Specifications

Chemical Identifiers

8007-43-0
C42H76O7
MFCD00151568
Arlacel™ 83; Span™ 83

Specifications

Yellow
Viscous Liquid
113°C (235°F)
C42H76O7
Arlacel™ 83; Span™ 83
693.06
8007-43-0
0.989
1.478
MFCD00151568
Insoluble in cold water,hot water. soluble in mineral oil and ethyl acetate.
Sorbitan sesquioleate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

missing translation for 'einecsNumber' : 232-360-1

missing translation for 'rtecsNumber' : WG2934330

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only