Learn More
Buffer Capsules, pH 4.00, Red, Reagecon™
Presented as an easy-to-use, color coded capsule. Reagecon™ Buffer Solution, pH 4.00, Red is tested at 25°C using an ISO 17025 accredited method and is NIST traceable. Each preservative-free capsule dissolves quickly into 100mL water to form a highly accurate calibration and control solution.
Brand: Thermo Scientific Russell CP1040
Code : 04
Additional Details : Weight : 0.12600kg
Description
These pH buffer capsules meet the same quality criteria as ready-to-use buffer solutions to ensure reliable, reportable pH measurements, along with the convenience and cost-savings of capsule form.
- Color coded for ease of identification
- Easy to use—dissolves quickly in 100mL distilled water
- Easy to store and transport with an extended shelf life
- Accuracy to ±0.02 pH units
- Traceable to SRM for NIST
- Certification to ISO 9001:2008
- Accredited to ISO 17025
Specifications
Red | |
1g/cm3 | |
Solid | |
Odorless | |
Miscibility with water: Soluble |
Yes | |
4.00 ±0.02 | |
50 Capsules | |
Yes |