missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Reduction reagent
£34.27 - £154.77
Chemical Identifiers
CAS | 10102-17-7 |
---|---|
Molecular Formula | Na2O3S2·5H2O |
Molecular Weight (g/mol) | 248.18 |
MDL Number | MFCD00149186 |
InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
PubChem CID | 61475 |
ChEBI | CHEBI:32150 |
IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10696052
|
Thermo Scientific Acros
202850010 |
1 kg | Plastic Bottle |
£34.27
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153930
|
Thermo Scientific Acros
202850025 |
2.5 kg | Plastic Bottle |
£68.24
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135163
|
Thermo Scientific Acros
202850100 |
10 kg | Plastic drum |
£154.77
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent for analysisChemical Identifiers
10102-17-7 | |
248.18 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Na2O3S2·5H2O | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Specifications
48.5°C | |
White | |
Crystals | |
Na2O3S2·5H2O | |
MFCD00149186 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.18 | |
Analytical | |
>45°C | |
Sodium thiosulfate pentahydrate |
10102-17-7 | |
6 to 8.4 (5% soln. at 25°C) | |
99.5 to 101% (Iodimetry) | |
Na2S2O3·5H2O | |
15, 8821 | |
Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in turpentine | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.18 | |
CHEBI:32150 | |
99.5% | |
1.72 | |
1.7200g/mL |
RUO – Research Use Only