missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Sulfide Nonahydrate, Honeywell Fluka™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Color | Colorless or Yellow |
Quantity | 100 g |
Linear Formula | Na2S·9H2O |
UN Number | 1849 |
Formula Weight | 240.18g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Chemical Name or Material | Sodium Sulfide Nonahydrate |
Safety and Handling
Product Identifier
- Sodium sulfide nonahydrate
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- EUH031-Contact with acids liberates toxic gas.
- EUH071-Corrosive to the respiratory tract.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium sulfide nonahydrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'