missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Nitrite, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 250 g |
Linear Formula | NaNO2 |
UN Number | 1500 |
Formula Weight | 69g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Ignition Point | 914°F |
Chemical Name or Material | Sodium Nitrite, Reag. Ph. Eur. |
Safety and Handling
Product Identifier
- Sodium nitrite
- Danger
- Acute toxicity Category 3
- ACUTE AQUATIC Acute 1
- Oxidising solid Category 3
- H272-May intensify fire; oxidiser.
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium nitrite
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction