missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitrate buffer solution (NaNO3) 5M (5N), SPEX CertiPrep™

Product Code. 11470209
Click to view available options
Quantity:
5 m
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Specifications

Quantity 5 m
compliance-icons
Product Identifier
  • 5M Sodium Nitrate Buffer
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium nitrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'