missing translation for 'onlineSavingsMsg'
Learn More
Sodium Iodide, puriss., Honeywell Fluka™ Product Code.: 15600680

Sodium Iodide, puriss., Honeywell Fluka™
BIOPHARMA

Product Code. 15600680 Shop All Honeywell Fluka Products
6 x 1 kg
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15600680

Brand: Honeywell-Fluka 031296X1KG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
MDL Number MFCD00003532
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 6 x 1 kg
Linear Formula NaI
UN Number UN3077
Formula Weight 149.89
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Density Vapor Density: >1 (vs air)
Chemical Name or Material Sodium Iodide, Meets analytical specification of Ph.Eur. BP, USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Iodide, puriss., Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.